Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGATTTATTCTCTACAATACTTC[A/G]TTAAAGTGTTCAATGATTTGCACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606017 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMMECR1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMMECR1L - AMMECR1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199140.1 | 5220 | UTR 3 | NP_001186069.1 | |||
NM_031445.2 | 5220 | Intron | NP_113633.2 | |||
XM_005263804.4 | 5220 | UTR 3 | XP_005263861.1 | |||
XM_005263806.4 | 5220 | UTR 3 | XP_005263863.1 | |||
XM_005263807.4 | 5220 | UTR 3 | XP_005263864.1 | |||
XM_011511952.2 | 5220 | UTR 3 | XP_011510254.1 | |||
XM_011511954.2 | 5220 | UTR 3 | XP_011510256.1 | |||
XM_011511955.2 | 5220 | Intron | XP_011510257.1 | |||
XM_011511956.2 | 5220 | Intron | XP_011510258.1 | |||
XM_017005063.1 | 5220 | Intron | XP_016860552.1 |
LOC107985803 - uncharacterized LOC107985803 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017005446.1 | 5220 | Intron | XP_016860935.1 |
POLR2D - RNA polymerase II subunit D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |