Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCATGTTCAAACTCTGAGAAGGA[C/T]AGCATGTTGTCATTGTCCAGATCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610646 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf70 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2orf70 - chromosome 2 open reading frame 70 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CIB4 - calcium and integrin binding family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029881.2 | 531 | Silent Mutation | CTA,CTG | L,L 159 | NP_001025052.1 | |
XM_011532514.2 | 531 | Silent Mutation | CTA,CTG | L,L 175 | XP_011530816.1 | |
XM_011532517.2 | 531 | Silent Mutation | CTA,CTG | L,L 161 | XP_011530819.1 | |
XM_017003329.1 | 531 | Silent Mutation | CTA,CTG | L,L 136 | XP_016858818.1 | |
XM_017003330.1 | 531 | Silent Mutation | CTA,CTG | L,L 114 | XP_016858819.1 | |
XM_017003331.1 | 531 | Intron | XP_016858820.1 |