Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACCTGTCGGCGGGGCTCCTGTCC[C/T]GGCTCGGCGCAGCCTTCAACTTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147556 | ||||||||||||||||||||
Literature Links: |
ITGA6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITGA6 - integrin subunit alpha 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000210.3 | 289 | Missense Mutation | CGG,TGG | R,W 19 | NP_000201.2 | |
NM_001079818.2 | 289 | Missense Mutation | CGG,TGG | R,W 19 | NP_001073286.1 | |
NM_001316306.1 | 289 | Intron | NP_001303235.1 | |||
XM_006712510.1 | 289 | Missense Mutation | CGG,TGG | R,W 19 | XP_006712573.1 | |
XM_006712511.1 | 289 | Missense Mutation | CGG,TGG | R,W 19 | XP_006712574.1 | |
XM_017004005.1 | 289 | Intron | XP_016859494.1 | |||
XM_017004006.1 | 289 | Intron | XP_016859495.1 | |||
XM_017004007.1 | 289 | Intron | XP_016859496.1 | |||
XM_017004008.1 | 289 | Intron | XP_016859497.1 |