Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612490 MIM: 616757 MIM: 611594 MIM: 607029 | ||||||||||||||||||||
Literature Links: |
C2orf68 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C2orf68 - chromosome 2 open reading frame 68 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF181 - ring finger protein 181 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM150A - transmembrane protein 150A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031738.2 | 1079 | Missense Mutation | TAT,TGT | Y,C 72 | NP_001026908.1 | |
XM_006711930.3 | 1079 | Missense Mutation | TAT,TGT | Y,C 72 | XP_006711993.1 | |
XM_006711931.3 | 1079 | Missense Mutation | TAT,TGT | Y,C 72 | XP_006711994.1 | |
XM_006711932.2 | 1079 | Intron | XP_006711995.1 | |||
XM_011532505.2 | 1079 | Missense Mutation | TAT,TGT | Y,C 48 | XP_011530807.1 | |
XM_011532507.2 | 1079 | Missense Mutation | TAT,TGT | Y,C 25 | XP_011530809.1 | |
XM_011532508.2 | 1079 | Intron | XP_011530810.1 | |||
XM_017003305.1 | 1079 | Missense Mutation | TAT,TGT | Y,C 25 | XP_016858794.1 | |
XM_017003306.1 | 1079 | Missense Mutation | TAT,TGT | Y,C 25 | XP_016858795.1 | |
XM_017003307.1 | 1079 | Missense Mutation | TAT,TGT | Y,C 25 | XP_016858796.1 |
USP39 - ubiquitin specific peptidase 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VAMP5 - vesicle associated membrane protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |