Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTTCCCTTATTCAGAGAAGATA[A/T]CAGACCAGGATTGCCAATGCTAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176871 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF2AK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF2AK2 - eukaryotic translation initiation factor 2 alpha kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPATCH11 - G-patch domain containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278505.1 | 201 | Intron | NP_001265434.1 | |||
NM_001322249.1 | 201 | Missense Mutation | AAC,ATC | N,I 45 | NP_001309178.1 | |
NM_174931.3 | 201 | Missense Mutation | AAC,ATC | N,I 45 | NP_777591.3 | |
XM_017003748.1 | 201 | Intron | XP_016859237.1 |
HEATR5B - HEAT repeat containing 5B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |