Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGCGAGCATCATCACTTTCATG[C/T]GTAATGTAACGTTCTCCAATGCGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615898 MIM: 607077 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFAF7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NDUFAF7 - NADH:ubiquinone oxidoreductase complex assembly factor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083946.1 | 3395 | Intron | NP_001077415.1 | |||
NM_144736.4 | 3395 | Intron | NP_653337.1 | |||
XM_005264410.1 | 3395 | Intron | XP_005264467.1 | |||
XM_005264411.1 | 3395 | Intron | XP_005264468.1 | |||
XM_011532956.1 | 3395 | Intron | XP_011531258.1 |
PRKD3 - protein kinase D3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005813.4 | 3395 | Silent Mutation | ACA,ACG | T,T 854 | NP_005804.1 | |
XM_005264237.3 | 3395 | Silent Mutation | ACA,ACG | T,T 854 | XP_005264294.1 | |
XM_005264238.3 | 3395 | Silent Mutation | ACA,ACG | T,T 750 | XP_005264295.1 | |
XM_017003741.1 | 3395 | Silent Mutation | ACA,ACG | T,T 681 | XP_016859230.1 | |
XM_017003742.1 | 3395 | Silent Mutation | ACA,ACG | T,T 681 | XP_016859231.1 | |
XM_017003743.1 | 3395 | Silent Mutation | ACA,ACG | T,T 595 | XP_016859232.1 | |
XM_017003744.1 | 3395 | Silent Mutation | ACA,ACG | T,T 595 | XP_016859233.1 |