Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTTCCCCGCCTACTGCATTTGAA[A/G]GATCCCGAAAACCCTGGATGATTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606247 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STAMBP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STAMBP - STAM binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006463.4 | 212 | Intron | NP_006454.1 | |||
NM_201647.2 | 212 | Intron | NP_964010.1 | |||
NM_213622.2 | 212 | UTR 5 | NP_998787.1 | |||
XM_005264088.3 | 212 | Intron | XP_005264145.1 | |||
XM_011532483.2 | 212 | Intron | XP_011530785.1 | |||
XM_011532484.2 | 212 | Intron | XP_011530786.1 | |||
XM_011532485.2 | 212 | Intron | XP_011530787.1 | |||
XM_017003175.1 | 212 | Intron | XP_016858664.1 | |||
XM_017003176.1 | 212 | Intron | XP_016858665.1 | |||
XM_017003177.1 | 212 | Intron | XP_016858666.1 | |||
XM_017003178.1 | 212 | UTR 5 | XP_016858667.1 | |||
XM_017003179.1 | 212 | Intron | XP_016858668.1 | |||
XM_017003180.1 | 212 | Intron | XP_016858669.1 | |||
XM_017003181.1 | 212 | UTR 5 | XP_016858670.1 |