Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGATCTCTAACAAACTTAATTG[C/T]CTGACAAAACGAAGTTAACTCTTTA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 602303 MIM: 609168 | |||||||||||||||||||||||
Literature Links: |
KAT2B PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
KAT2B - lysine acetyltransferase 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SGO1 - shugoshin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012409.3 | Intron | NP_001012409.1 | ||||
NM_001012410.4 | Intron | NP_001012410.1 | ||||
NM_001012411.3 | Intron | NP_001012411.1 | ||||
NM_001012412.4 | Intron | NP_001012412.1 | ||||
NM_001012413.3 | Intron | NP_001012413.1 | ||||
NM_001199251.2 | Intron | NP_001186180.1 | ||||
NM_001199252.2 | Intron | NP_001186181.1 | ||||
NM_001199253.2 | Intron | NP_001186182.1 | ||||
NM_001199254.2 | Intron | NP_001186183.1 | ||||
NM_001199255.2 | Intron | NP_001186184.1 | ||||
NM_001199256.2 | Intron | NP_001186185.1 | ||||
NM_001199257.2 | Intron | NP_001186186.1 | ||||
NM_138484.4 | Intron | NP_612493.1 | ||||
XM_011533373.2 | Intron | XP_011531675.1 | ||||
XM_011533375.2 | Intron | XP_011531677.1 | ||||
XM_011533376.2 | Intron | XP_011531678.1 | ||||
XM_011533377.2 | Intron | XP_011531679.1 |