Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCGTTGTTCATTTCAGCAGGGAG[C/T]CAGGAAACTGGACATTTCGGGAGGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 608708 | |||||||||||||||||||||||
Literature Links: |
BOC PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
BOC - BOC cell adhesion associated, oncogene regulated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301861.1 | Intron | NP_001288790.1 | ||||
NM_001301867.1 | Intron | NP_001288796.1 | ||||
NM_033254.3 | Intron | NP_150279.1 | ||||
XM_005247891.2 | Intron | XP_005247948.1 | ||||
XM_005247892.2 | Intron | XP_005247949.1 | ||||
XM_005247897.2 | Intron | XP_005247954.1 | ||||
XM_011513305.2 | Intron | XP_011511607.1 | ||||
XM_017007441.1 | Intron | XP_016862930.1 | ||||
XM_017007442.1 | Intron | XP_016862931.1 | ||||
XM_017007443.1 | Intron | XP_016862932.1 | ||||
XM_017007444.1 | Intron | XP_016862933.1 | ||||
XM_017007445.1 | Intron | XP_016862934.1 | ||||
XM_017007446.1 | Intron | XP_016862935.1 | ||||
XM_017007447.1 | Intron | XP_016862936.1 | ||||
XM_017007448.1 | Intron | XP_016862937.1 | ||||
XM_017007449.1 | Intron | XP_016862938.1 | ||||
XM_017007450.1 | Intron | XP_016862939.1 | ||||
XM_017007451.1 | Intron | XP_016862940.1 | ||||
XM_017007452.1 | Intron | XP_016862941.1 | ||||
XM_017007453.1 | Intron | XP_016862942.1 | ||||
XM_017007454.1 | Intron | XP_016862943.1 | ||||
XM_017007455.1 | Intron | XP_016862944.1 |