Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATACCTTTATAGAGAAAATTCCAC[C/T]TCAGTTTGCACAGGCTATAGCCAAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 616607 | |||||||||||||||||||||||
Literature Links: |
KBTBD8 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
KBTBD8 - kelch repeat and BTB domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032505.2 | 602 | Missense Mutation | CCT,CTT | P,L 255 | NP_115894.2 | |
XM_005264771.3 | 602 | Missense Mutation | CCT,CTT | P,L 178 | XP_005264828.1 |