Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGGAGAGGTCGCGGGCTGGGAGC[C/T]ACATCTCCTCCAATCGAGCGTATGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609910 | |||||||||||||||||||||||
Literature Links: |
MAATS1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
MAATS1 - MYCBP associated and testis expressed 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320316.1 | 165 | UTR 5 | NP_001307245.1 | |||
NM_001320317.1 | 165 | UTR 5 | NP_001307246.1 | |||
NM_001320318.1 | 165 | UTR 5 | NP_001307247.1 | |||
NM_033364.3 | 165 | Missense Mutation | CAC,TAC | H,Y 30 | NP_203528.2 | |
XM_011513277.1 | 165 | Intron | XP_011511579.1 |