Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGCATTGAGATCAATATCATCAC[A/G]GAAGGATGGCCGGCGGAGCTTCAGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 605958 | |||||||||||||||||||||||
Literature Links: |
TRAIP PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TRAIP - TRAF interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005879.2 | 765 | Missense Mutation | CGT,TGT | R,C 311 | NP_005870.2 | |
XM_011533264.1 | 765 | Missense Mutation | CGT,TGT | R,C 234 | XP_011531566.1 | |
XM_017005526.1 | 765 | Missense Mutation | CGT,TGT | R,C 212 | XP_016861015.1 |