Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTTGCCTATATTCAGTACGAGA[C/G]CTTGGACGCAGCCCAGGCCGCCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603123 MIM: 601916 MIM: 612602 | ||||||||||||||||||||
Literature Links: |
DCAF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCAF1 - DDB1 and CUL4 associated factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DOCK3 - dedicator of cytokinesis 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MANF - mesencephalic astrocyte derived neurotrophic factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM15B - RNA binding motif protein 15B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013286.4 | 1517 | Missense Mutation | ACC,AGC | T,S 462 | NP_037418.3 |