Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTTGTCACTGACATACAGGGCC[G/T]TGTTGATGTCCCTGGTGAAGACACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600249 MIM: 610803 | ||||||||||||||||||||
Literature Links: |
ALDH1L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALDH1L1 - aldehyde dehydrogenase 1 family member L1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270364.1 | 2785 | Missense Mutation | AAG,ACG | K,T 855 | NP_001257293.1 | |
NM_001270365.1 | 2785 | Missense Mutation | AAG,ACG | K,T 744 | NP_001257294.1 | |
NM_012190.3 | 2785 | Missense Mutation | AAG,ACG | K,T 845 | NP_036322.2 | |
XM_006713481.2 | 2785 | Missense Mutation | AAG,ACG | K,T 845 | XP_006713544.1 | |
XM_011512355.1 | 2785 | Missense Mutation | AAG,ACG | K,T 845 | XP_011510657.1 | |
XM_017005613.1 | 2785 | Missense Mutation | AAG,ACG | K,T 781 | XP_016861102.1 | |
XM_017005614.1 | 2785 | Missense Mutation | AAG,ACG | K,T 771 | XP_016861103.1 |
ALDH1L1-AS1 - ALDH1L1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC41A3 - solute carrier family 41 member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |