Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCGGGCAGCATGAGCGGCTGCGG[G/T]CTCTTCCTGCGCACCACGGCTGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611103 | ||||||||||||||||||||
Literature Links: |
ACAD9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACAD9 - acyl-CoA dehydrogenase family member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014049.4 | 217 | Silent Mutation | GGG,GGT | G,G 5 | NP_054768.2 | |
XM_017006264.1 | 217 | UTR 5 | XP_016861753.1 | |||
XM_017006265.1 | 217 | Intron | XP_016861754.1 |
LOC653712 - intraflagellar transport 122 homolog (Chlamydomonas) pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |