Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGGACTGAAGCCCTATACATGC[A/G]GTGAATGTGGGAAATCTTTCAGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF660 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF660 - zinc finger protein 660 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173658.2 | 658 | Missense Mutation | AGT,GGT | S,G 109 | NP_775929.2 | |
XM_005265090.4 | 658 | Missense Mutation | AGT,GGT | S,G 109 | XP_005265147.1 | |
XM_006713117.3 | 658 | Missense Mutation | AGT,GGT | S,G 109 | XP_006713180.1 | |
XM_011533622.2 | 658 | Missense Mutation | AGT,GGT | S,G 109 | XP_011531924.1 |