Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTAGATCTTTCCAATGAACACTA[A/G]TAGAGTACTCTGCTCTTGGCTGGAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
FAM19A1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
FAM19A1 - family with sequence similarity 19 member A1, C-C motif chemokine like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252216.1 | 890 | Intron | NP_001239145.1 | |||
NM_213609.3 | 890 | UTR 5 | NP_998774.2 | |||
XM_017006386.1 | 890 | Intron | XP_016861875.1 | |||
XM_017006387.1 | 890 | UTR 5 | XP_016861876.1 | |||
XM_017006388.1 | 890 | UTR 5 | XP_016861877.1 | |||
XM_017006389.1 | 890 | Intron | XP_016861878.1 | |||
XM_017006390.1 | 890 | UTR 5 | XP_016861879.1 |