Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGAAACCTTCACCTAAATGATAA[C/T]ATCTTAACGGTCACGCATATGAAAC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609729 | |||||||||||||||||||||||
Literature Links: |
PDZRN3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PDZRN3 - PDZ domain containing ring finger 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303139.1 | 3112 | UTR 3 | NP_001290068.1 | |||
NM_001303140.1 | 3112 | UTR 3 | NP_001290069.1 | |||
NM_001303141.1 | 3112 | UTR 3 | NP_001290070.1 | |||
NM_001303142.1 | 3112 | UTR 3 | NP_001290071.1 | |||
NM_015009.2 | 3112 | UTR 3 | NP_055824.1 | |||
XM_017005942.1 | 3112 | UTR 3 | XP_016861431.1 |