Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTTACAAAGATGAGCGGAACTG[C/G]CAGTTCTGCCACTGCACCTGTTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TRIM42 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIM42 - tripartite motif containing 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152616.4 | 356 | Missense Mutation | TGC,TGG | C,W 50 | NP_689829.3 | |
XM_011512740.2 | 356 | Nonsense Mutation | TGC,TGG | C,W 50 | XP_011511042.1 |