Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAATTCCCAGGTAGGGGTCTCTG[A/G]TAGTAATGGAACTCTATAAACTATT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
CCDC191 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CCDC191 - coiled-coil domain containing 191 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
QTRT2 - queuine tRNA-ribosyltransferase accessory subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256835.1 | Intron | NP_001243764.1 | ||||
NM_001256836.1 | Intron | NP_001243765.1 | ||||
NM_001256837.1 | Intron | NP_001243766.1 | ||||
NM_024638.3 | Intron | NP_078914.1 | ||||
XM_005247769.3 | Intron | XP_005247826.1 | ||||
XM_005247770.3 | Intron | XP_005247827.1 | ||||
XM_006713744.3 | Intron | XP_006713807.1 | ||||
XM_011513137.2 | Intron | XP_011511439.1 | ||||
XM_017007182.1 | Intron | XP_016862671.1 | ||||
XM_017007183.1 | Intron | XP_016862672.1 | ||||
XM_017007184.1 | Intron | XP_016862673.1 |