Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTGTAAATATATTGGACCATTT[G/T]TTTGTCGTTCTTTGTGTTTTCTTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603598 | ||||||||||||||||||||
Literature Links: |
TNFSF10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TNFSF10 - tumor necrosis factor superfamily member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190942.1 | 736 | UTR 3 | NP_001177871.1 | |||
NM_001190943.1 | 736 | Intron | NP_001177872.1 | |||
NM_003810.3 | 736 | Missense Mutation | AAA,CAA | K,Q 205 | NP_003801.1 |