Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCGCATCTCCAAACTCCTCATG[A/G]TCTGCCAGAATGCAGGCATTTCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616735 | ||||||||||||||||||||
Literature Links: |
C3orf20 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C3orf20 - chromosome 3 open reading frame 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184957.1 | 543 | Intron | NP_001171886.1 | |||
NM_001184958.1 | 543 | Intron | NP_001171887.1 | |||
NM_032137.4 | 543 | Missense Mutation | ATC,GTC | I,V 31 | NP_115513.4 | |
XM_006713341.2 | 543 | Missense Mutation | ATC,GTC | I,V 31 | XP_006713404.1 | |
XM_006713342.2 | 543 | Missense Mutation | ATC,GTC | I,V 31 | XP_006713405.1 | |
XM_006713343.2 | 543 | Missense Mutation | ATC,GTC | I,V 31 | XP_006713406.1 | |
XM_011534153.2 | 543 | Missense Mutation | ATC,GTC | I,V 31 | XP_011532455.1 | |
XM_011534154.2 | 543 | Intron | XP_011532456.1 | |||
XM_011534157.2 | 543 | Intron | XP_011532459.1 | |||
XM_011534158.2 | 543 | Missense Mutation | ATC,GTC | I,V 31 | XP_011532460.1 | |
XM_017007310.1 | 543 | Missense Mutation | ATC,GTC | I,V 31 | XP_016862799.1 |
CCDC174 - coiled-coil domain containing 174 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |