Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCTGGAGTTTCTTGACCAAAGA[A/G]CTTGGTAGTCATTCCCTTCAAAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605931 | ||||||||||||||||||||
Literature Links: |
SNX4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SNX4 - sorting nexin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003794.3 | 1214 | Missense Mutation | CTC,TTC | L,F 362 | NP_003785.1 | |
XM_017007414.1 | 1214 | Missense Mutation | CTC,TTC | L,F 394 | XP_016862903.1 |