Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCACTCAATCCGGATCGAGGGGGA[C/T]GTCACCCTCGGGGGGCTGTTCCCCG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604101 | |||||||||||||||||||||||
Literature Links: |
GRM7 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GRM7 - glutamate metabotropic receptor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000844.3 | 418 | Silent Mutation | GAC,GAT | D,D 48 | NP_000835.1 | |
NM_181874.2 | 418 | Silent Mutation | GAC,GAT | D,D 48 | NP_870989.1 | |
XM_017006272.1 | 418 | Intron | XP_016861761.1 | |||
XM_017006273.1 | 418 | Intron | XP_016861762.1 |