Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTATATTCACTGCTACAACAGGA[C/G]CAGTAACAACTATATTTATTTAAAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607911 MIM: 120436 | |||||||||||||||||||||||
Literature Links: |
EPM2AIP1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
EPM2AIP1 - EPM2A interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014805.3 | 4414 | UTR 3 | NP_055620.1 |
MLH1 - mutL homolog 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |