Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAAGCTTAAATTTGTATTCGCTA[A/G]TTTGTCTATTGGCATCTCCTAGAAC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 614043 MIM: 120436 | |||||||||||||||||||||||
Literature Links: |
LRRFIP2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LRRFIP2 - LRR binding FLII interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134369.2 | 1685 | Missense Mutation | ACT,ATT | T,I 333 | NP_001127841.1 | |
NM_001282691.1 | 1685 | Missense Mutation | ACT,ATT | T,I 412 | NP_001269620.1 | |
NM_006309.3 | 1685 | Missense Mutation | ACT,ATT | T,I 630 | NP_006300.1 | |
NM_017724.2 | 1685 | Missense Mutation | ACT,ATT | T,I 309 | NP_060194.1 | |
XM_005265539.2 | 1685 | Missense Mutation | ACT,ATT | T,I 630 | XP_005265596.1 | |
XM_005265540.1 | 1685 | Missense Mutation | ACT,ATT | T,I 630 | XP_005265597.1 | |
XM_005265550.1 | 1685 | Missense Mutation | ACT,ATT | T,I 486 | XP_005265607.1 | |
XM_005265551.1 | 1685 | Missense Mutation | ACT,ATT | T,I 449 | XP_005265608.1 | |
XM_005265553.1 | 1685 | Missense Mutation | ACT,ATT | T,I 398 | XP_005265610.1 | |
XM_005265554.2 | 1685 | Missense Mutation | ACT,ATT | T,I 398 | XP_005265611.1 | |
XM_005265556.1 | 1685 | Missense Mutation | ACT,ATT | T,I 374 | XP_005265613.1 | |
XM_006713385.1 | 1685 | Missense Mutation | ACT,ATT | T,I 629 | XP_006713448.1 | |
XM_006713386.1 | 1685 | Missense Mutation | ACT,ATT | T,I 616 | XP_006713449.1 | |
XM_006713387.1 | 1685 | Missense Mutation | ACT,ATT | T,I 615 | XP_006713450.1 | |
XM_006713388.1 | 1685 | Missense Mutation | ACT,ATT | T,I 614 | XP_006713451.1 | |
XM_006713389.1 | 1685 | Missense Mutation | ACT,ATT | T,I 613 | XP_006713452.1 | |
XM_006713390.1 | 1685 | Missense Mutation | ACT,ATT | T,I 613 | XP_006713453.1 | |
XM_006713392.1 | 1685 | Missense Mutation | ACT,ATT | T,I 606 | XP_006713455.1 | |
XM_006713393.1 | 1685 | Missense Mutation | ACT,ATT | T,I 600 | XP_006713456.1 | |
XM_006713394.1 | 1685 | Missense Mutation | ACT,ATT | T,I 599 | XP_006713457.1 | |
XM_006713395.1 | 1685 | Missense Mutation | ACT,ATT | T,I 599 | XP_006713458.1 | |
XM_006713396.1 | 1685 | Missense Mutation | ACT,ATT | T,I 594 | XP_006713459.1 | |
XM_006713397.1 | 1685 | Missense Mutation | ACT,ATT | T,I 558 | XP_006713460.1 | |
XM_006713400.1 | 1685 | Missense Mutation | ACT,ATT | T,I 413 | XP_006713463.1 | |
XM_006713404.1 | 1685 | Missense Mutation | ACT,ATT | T,I 324 | XP_006713467.1 | |
XM_011534217.1 | 1685 | Missense Mutation | ACT,ATT | T,I 630 | XP_011532519.1 | |
XM_011534218.1 | 1685 | Missense Mutation | ACT,ATT | T,I 630 | XP_011532520.1 | |
XM_011534219.1 | 1685 | Missense Mutation | ACT,ATT | T,I 630 | XP_011532521.1 | |
XM_011534221.1 | 1685 | Missense Mutation | ACT,ATT | T,I 594 | XP_011532523.1 | |
XM_011534222.1 | 1685 | Missense Mutation | ACT,ATT | T,I 565 | XP_011532524.1 | |
XM_011534229.1 | 1685 | Missense Mutation | ACT,ATT | T,I 413 | XP_011532531.1 | |
XM_017007465.1 | 1685 | Missense Mutation | ACT,ATT | T,I 579 | XP_016862954.1 | |
XM_017007466.1 | 1685 | Missense Mutation | ACT,ATT | T,I 577 | XP_016862955.1 | |
XM_017007467.1 | 1685 | Missense Mutation | ACT,ATT | T,I 575 | XP_016862956.1 | |
XM_017007468.1 | 1685 | Missense Mutation | ACT,ATT | T,I 543 | XP_016862957.1 | |
XM_017007469.1 | 1685 | Missense Mutation | ACT,ATT | T,I 541 | XP_016862958.1 | |
XM_017007470.1 | 1685 | Missense Mutation | ACT,ATT | T,I 529 | XP_016862959.1 | |
XM_017007471.1 | 1685 | Missense Mutation | ACT,ATT | T,I 432 | XP_016862960.1 | |
XM_017007472.1 | 1685 | Missense Mutation | ACT,ATT | T,I 418 | XP_016862961.1 | |
XM_017007473.1 | 1685 | Missense Mutation | ACT,ATT | T,I 408 | XP_016862962.1 | |
XM_017007474.1 | 1685 | Missense Mutation | ACT,ATT | T,I 401 | XP_016862963.1 | |
XM_017007475.1 | 1685 | Missense Mutation | ACT,ATT | T,I 401 | XP_016862964.1 | |
XM_017007476.1 | 1685 | Missense Mutation | ACT,ATT | T,I 398 | XP_016862965.1 | |
XM_017007477.1 | 1685 | Missense Mutation | ACT,ATT | T,I 377 | XP_016862966.1 | |
XM_017007478.1 | 1685 | Missense Mutation | ACT,ATT | T,I 367 | XP_016862967.1 | |
XM_017007479.1 | 1685 | Missense Mutation | ACT,ATT | T,I 367 | XP_016862968.1 | |
XM_017007480.1 | 1685 | Missense Mutation | ACT,ATT | T,I 367 | XP_016862969.1 | |
XM_017007481.1 | 1685 | Missense Mutation | ACT,ATT | T,I 364 | XP_016862970.1 | |
XM_017007482.1 | 1685 | Missense Mutation | ACT,ATT | T,I 343 | XP_016862971.1 | |
XM_017007483.1 | 1685 | Missense Mutation | ACT,ATT | T,I 333 | XP_016862972.1 | |
XM_017007484.1 | 1685 | Missense Mutation | ACT,ATT | T,I 333 | XP_016862973.1 |
MLH1 - mutL homolog 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |