Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCCTGTCATTCTGCCCTTGAAT[A/G]TTACACACACAAAAAAATCAAGCTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 614789 MIM: 601126 | |||||||||||||||||||||||
Literature Links: |
EOGT PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
EOGT - EGF domain specific O-linked N-acetylglucosamine transferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMF1 - TATA element modulatory factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007114.2 | 5306 | UTR 3 | NP_009045.2 | |||
XM_011534056.2 | 5306 | UTR 3 | XP_011532358.1 |