Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGCCTAGATATCTGGTGAGTTG[A/G]GCCCGCCCTTCTCTTTTCTGAAATG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 616702 | |||||||||||||||||||||||
Literature Links: |
ZNF589 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ZNF589 - zinc finger protein 589 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016089.2 | Intron | NP_057173.2 | ||||
XM_017006575.1 | Intron | XP_016862064.1 | ||||
XM_017006576.1 | Intron | XP_016862065.1 | ||||
XM_017006577.1 | Intron | XP_016862066.1 | ||||
XM_017006578.1 | Intron | XP_016862067.1 | ||||
XM_017006579.1 | Intron | XP_016862068.1 |