Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGCTACAGGGACCAGATCAGGG[A/C]CAAAGGGACTTGCTGCCTCCGGTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608263 MIM: 610028 | ||||||||||||||||||||
Literature Links: |
HSPBAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HSPBAP1 - HSPB1 associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320728.1 | 767 | Missense Mutation | GGC,GTC | G,V 361 | NP_001307657.1 | |
NM_024610.5 | 767 | Missense Mutation | GGC,GTC | G,V 389 | NP_078886.2 | |
XM_005247762.2 | 767 | Intron | XP_005247819.1 | |||
XM_011513130.2 | 767 | Missense Mutation | GGC,GTC | G,V 352 | XP_011511432.1 | |
XM_011513132.2 | 767 | Missense Mutation | GGC,GTC | G,V 350 | XP_011511434.2 | |
XM_017007179.1 | 767 | Missense Mutation | GGC,GTC | G,V 327 | XP_016862668.1 | |
XM_017007180.1 | 767 | Missense Mutation | GGC,GTC | G,V 154 | XP_016862669.1 |
PARP14 - poly(ADP-ribose) polymerase family member 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |