Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCGTGCTGGACTGCTCTGGTGC[C/T]CGGAGCCGTTTCATGAGGGTGGTCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 614993 MIM: 605958 | |||||||||||||||||||||||
Literature Links: |
CAMKV PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CAMKV - CaM kinase like vesicle associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320147.1 | 1460 | Silent Mutation | CGA,CGG | R,R 327 | NP_001307076.1 | |
NM_024046.4 | 1460 | Silent Mutation | CGA,CGG | R,R 327 | NP_076951.2 |
TRAIP - TRAF interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |