Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGTTTTGGAAACTAAAACACCA[A/G]ATAAAAATTACCATTCCTAATCTAC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
ARMC8 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ARMC8 - armadillo repeat containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267041.2 | 1208 | Intron | NP_001253970.1 | |||
NM_001267042.2 | 1208 | Intron | NP_001253971.1 | |||
NM_001282342.1 | 1208 | Intron | NP_001269271.1 | |||
NM_014154.4 | 1208 | Intron | NP_054873.2 | |||
NM_015396.5 | 1208 | Intron | NP_056211.2 | |||
NM_213654.3 | 1208 | Intron | NP_998819.1 | |||
XM_006713564.3 | 1208 | Intron | XP_006713627.1 | |||
XM_006713565.2 | 1208 | Intron | XP_006713628.1 | |||
XM_006713566.3 | 1208 | Intron | XP_006713629.1 | |||
XM_006713567.3 | 1208 | Intron | XP_006713630.1 | |||
XM_011512632.1 | 1208 | Intron | XP_011510934.1 | |||
XM_011512633.1 | 1208 | Intron | XP_011510935.1 | |||
XM_017006095.1 | 1208 | Intron | XP_016861584.1 | |||
XM_017006096.1 | 1208 | Intron | XP_016861585.1 | |||
XM_017006097.1 | 1208 | Intron | XP_016861586.1 | |||
XM_017006098.1 | 1208 | Intron | XP_016861587.1 | |||
XM_017006099.1 | 1208 | Intron | XP_016861588.1 | |||
XM_017006100.1 | 1208 | Intron | XP_016861589.1 | |||
XM_017006101.1 | 1208 | Intron | XP_016861590.1 |
NME9 - NME/NM23 family member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178130.3 | 1208 | UTR 3 | NP_835231.1 | |||
XM_005247429.3 | 1208 | Intron | XP_005247486.1 | |||
XM_006713615.3 | 1208 | Intron | XP_006713678.1 | |||
XM_011512774.2 | 1208 | Intron | XP_011511076.1 | |||
XM_011512777.2 | 1208 | Intron | XP_011511079.1 | |||
XM_017006306.1 | 1208 | Intron | XP_016861795.1 | |||
XM_017006307.1 | 1208 | Intron | XP_016861796.1 | |||
XM_017006308.1 | 1208 | Intron | XP_016861797.1 | |||
XM_017006309.1 | 1208 | Intron | XP_016861798.1 | |||
XM_017006310.1 | 1208 | Intron | XP_016861799.1 | |||
XM_017006311.1 | 1208 | Intron | XP_016861800.1 | |||
XM_017006312.1 | 1208 | Intron | XP_016861801.1 | |||
XM_017006313.1 | 1208 | Intron | XP_016861802.1 | |||
XM_017006314.1 | 1208 | Intron | XP_016861803.1 | |||
XM_017006315.1 | 1208 | Intron | XP_016861804.1 | |||
XM_017006316.1 | 1208 | Intron | XP_016861805.1 | |||
XM_017006317.1 | 1208 | Intron | XP_016861806.1 | |||
XM_017006318.1 | 1208 | Intron | XP_016861807.1 | |||
XM_017006319.1 | 1208 | Intron | XP_016861808.1 |