Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGGCCTGTGGGGTTTCATTAAT[A/G]GAACACAGTCTTTCTTCTGGTCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601802 | ||||||||||||||||||||
Literature Links: |
HESX1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HESX1 - HESX homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003865.2 | 338 | Missense Mutation | CCA,CTA | P,L 37 | NP_003856.1 | |
XM_005265526.4 | 338 | Missense Mutation | CCA,CTA | P,L 37 | XP_005265583.1 | |
XM_006713379.3 | 338 | Missense Mutation | CCA,CTA | P,L 37 | XP_006713442.1 | |
XM_011534204.2 | 338 | Missense Mutation | CCA,CTA | P,L 37 | XP_011532506.1 | |
XM_011534205.2 | 338 | Missense Mutation | CCA,CTA | P,L 37 | XP_011532507.1 | |
XM_017007421.1 | 338 | Missense Mutation | CCA,CTA | P,L 37 | XP_016862910.1 |