Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTGAAACAGCACAGGACCACCA[A/G]GAGGCTGGTCGTGCTCGCCACCCCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 603942 | |||||||||||||||||||||||
Literature Links: |
GYG1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GYG1 - glycogenin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184720.1 | 331 | Missense Mutation | AAG,AGG | K,R 33 | NP_001171649.1 | |
NM_001184721.1 | 331 | Missense Mutation | AAG,AGG | K,R 33 | NP_001171650.1 | |
NM_004130.3 | 331 | Missense Mutation | AAG,AGG | K,R 33 | NP_004121.2 | |
XM_017006275.1 | 331 | Intron | XP_016861764.1 | |||
XM_017006276.1 | 331 | Intron | XP_016861765.1 |