Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTTTTGCAAAAGAAGCCAGTTG[A/G]TTCAGAAACATCACAGGCAAAAGGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
CCDC66 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CCDC66 - coiled-coil domain containing 66 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012506.4 | 289 | Missense Mutation | GAT,GGT | D,G 40 | NP_001012524.4 | |
NM_001141947.1 | 289 | Missense Mutation | GAT,GGT | D,G 74 | NP_001135419.1 | |
XM_005265081.3 | 289 | Missense Mutation | GAT,GGT | D,G 74 | XP_005265138.1 | |
XM_005265082.3 | 289 | Missense Mutation | GAT,GGT | D,G 74 | XP_005265139.1 | |
XM_005265083.3 | 289 | Missense Mutation | GAT,GGT | D,G 74 | XP_005265140.1 | |
XM_005265084.3 | 289 | UTR 5 | XP_005265141.1 | |||
XM_011533614.2 | 289 | Missense Mutation | GAT,GGT | D,G 41 | XP_011531916.1 | |
XM_011533615.2 | 289 | Missense Mutation | GAT,GGT | D,G 41 | XP_011531917.1 | |
XM_011533616.2 | 289 | Missense Mutation | GAT,GGT | D,G 40 | XP_011531918.1 | |
XM_011533617.2 | 289 | Missense Mutation | GAT,GGT | D,G 40 | XP_011531919.1 | |
XM_011533619.2 | 289 | UTR 5 | XP_011531921.1 | |||
XM_017006229.1 | 289 | Missense Mutation | GAT,GGT | D,G 74 | XP_016861718.1 | |
XM_017006230.1 | 289 | Missense Mutation | GAT,GGT | D,G 41 | XP_016861719.1 | |
XM_017006231.1 | 289 | Missense Mutation | GAT,GGT | D,G 41 | XP_016861720.1 | |
XM_017006232.1 | 289 | Missense Mutation | GAT,GGT | D,G 74 | XP_016861721.1 | |
XM_017006233.1 | 289 | Missense Mutation | GAT,GGT | D,G 74 | XP_016861722.1 | |
XM_017006234.1 | 289 | Missense Mutation | GAT,GGT | D,G 40 | XP_016861723.1 | |
XM_017006235.1 | 289 | Missense Mutation | GAT,GGT | D,G 40 | XP_016861724.1 | |
XM_017006236.1 | 289 | UTR 5 | XP_016861725.1 | |||
XM_017006237.1 | 289 | UTR 5 | XP_016861726.1 | |||
XM_017006238.1 | 289 | UTR 5 | XP_016861727.1 | |||
XM_017006239.1 | 289 | Intron | XP_016861728.1 | |||
XM_017006240.1 | 289 | UTR 5 | XP_016861729.1 | |||
XM_017006241.1 | 289 | UTR 5 | XP_016861730.1 | |||
XM_017006242.1 | 289 | UTR 5 | XP_016861731.1 | |||
XM_017006243.1 | 289 | UTR 5 | XP_016861732.1 | |||
XM_017006244.1 | 289 | UTR 5 | XP_016861733.1 | |||
XM_017006245.1 | 289 | UTR 5 | XP_016861734.1 |