Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTAGAAAAGAAGTCTATCAAAGG[A/G]ATACGAAAAGCTGTCTCCACCTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR5H6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR5H6 - olfactory receptor family 5 subfamily H member 6 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005479.1 | 793 | Silent Mutation | GGA,GGG | G,G 249 | NP_001005479.1 | |
XM_011513118.2 | 793 | Silent Mutation | GGA,GGG | G,G 233 | XP_011511420.1 |