Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCCCTGACTGACAGGATCAGAA[C/T]GTTGACCAGCCGAGTCATCGCCTCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
TRANK1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TRANK1 - tetratricopeptide repeat and ankyrin repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014831.2 | 9785 | Missense Mutation | NP_055646.2 | |||
XM_006713432.3 | 9785 | Missense Mutation | XP_006713495.1 | |||
XM_011534289.2 | 9785 | Missense Mutation | XP_011532591.1 | |||
XM_017007570.1 | 9785 | Missense Mutation | XP_016863059.1 | |||
XM_017007571.1 | 9785 | Missense Mutation | XP_016863060.1 | |||
XM_017007572.1 | 9785 | Missense Mutation | XP_016863061.1 | |||
XM_017007573.1 | 9785 | Intron | XP_016863062.1 |