Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGTTTCCTTCTAGCATTTGAGAC[A/G]GAGTTGGCTTATCTCTCAGGAAATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609075 | ||||||||||||||||||||
Literature Links: |
FBXW12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXW12 - F-box and WD repeat domain containing 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159927.1 | 309 | Silent Mutation | ACA,ACG | T,T 99 | NP_001153399.1 | |
NM_001159929.1 | 309 | Silent Mutation | ACA,ACG | T,T 80 | NP_001153401.1 | |
NM_207102.2 | 309 | Silent Mutation | ACA,ACG | T,T 99 | NP_996985.2 | |
XM_017006224.1 | 309 | Silent Mutation | ACA,ACG | T,T 99 | XP_016861713.1 |