Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCCAAACGAGCAGTCCTGTGTTC[A/C]GGCGAGGACAGGTGTTTCACCTGCG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 600585 | |||||||||||||||||||||||
Literature Links: |
TGM4 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TGM4 - transglutaminase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003241.3 | 182 | Silent Mutation | AGG,CGG | R,R 37 | NP_003232.2 | |
XM_011534042.1 | 182 | Silent Mutation | AGG,CGG | R,R 82 | XP_011532344.1 |