Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGGAAGGTGGACCCACCTGTGC[A/G]GCACCTGGAATTCAGAGGAAGGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602492 MIM: 609594 | ||||||||||||||||||||
Literature Links: |
PTX3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTX3 - pentraxin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002852.3 | 957 | Missense Mutation | AGC,GGC | S,G 272 | NP_002843.2 |
VEPH1 - ventricular zone expressed PH domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167911.1 | 957 | Intron | NP_001161383.1 | |||
NM_001167912.1 | 957 | Intron | NP_001161384.1 | |||
NM_001167915.1 | 957 | Intron | NP_001161387.1 | |||
NM_001167916.1 | 957 | Intron | NP_001161388.1 | |||
NM_001167917.1 | 957 | Intron | NP_001161389.1 | |||
NM_024621.2 | 957 | Intron | NP_078897.2 | |||
XM_011513134.2 | 957 | Intron | XP_011511436.1 | |||
XM_011513135.1 | 957 | Intron | XP_011511437.1 |