Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACGATGGAGGAGCCGCAGCGCGC[C/G]CGCTCGCACACAGTCACCACCACCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607891 | ||||||||||||||||||||
Literature Links: |
CMTM8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CMTM8 - CKLF like MARVEL transmembrane domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320308.1 | 46 | Silent Mutation | GCC,GCG | A,A 7 | NP_001307237.1 | |
NM_178868.4 | 46 | Silent Mutation | GCC,GCG | A,A 7 | NP_849199.2 | |
XM_011533416.2 | 46 | Silent Mutation | GCC,GCG | A,A 7 | XP_011531718.1 | |
XM_017005779.1 | 46 | Intron | XP_016861268.1 | |||
XM_017005780.1 | 46 | Intron | XP_016861269.1 |