Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAAGCAAGAGAAAAAAGAAACAC[C/T]GTAGACGGTCCTCCTCGAGCAGTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613352 | ||||||||||||||||||||
Literature Links: |
RSRC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RSRC1 - arginine and serine rich coiled-coil 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271834.1 | 235 | Missense Mutation | CGT,TGT | R,C 21 | NP_001258763.1 | |
NM_001271838.1 | 235 | Missense Mutation | CGT,TGT | R,C 21 | NP_001258767.1 | |
NM_016625.3 | 235 | Missense Mutation | CGT,TGT | R,C 21 | NP_057709.2 |