Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGCCCTCCCGCCGGGCCACAGC[A/G]TAGGGCTGGGGTAGGTGTACATCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608336 | ||||||||||||||||||||
Literature Links: |
MIR6825 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR6825 - microRNA 6825 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPRA1 - transmembrane protein adipocyte associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136053.2 | 1337 | Silent Mutation | TAC,TAT | Y,Y 312 | NP_001129525.1 | |
NM_001142646.2 | 1337 | Missense Mutation | CGC,TGC | R,C 258 | NP_001136118.1 | |
XM_006713495.3 | 1337 | Silent Mutation | TAC,TAT | Y,Y 312 | XP_006713558.1 | |
XM_006713496.2 | 1337 | Silent Mutation | TAC,TAT | Y,Y 312 | XP_006713559.1 | |
XM_006713497.3 | 1337 | Silent Mutation | TAC,TAT | Y,Y 312 | XP_006713560.1 | |
XM_011512422.1 | 1337 | Silent Mutation | TAC,TAT | Y,Y 312 | XP_011510724.1 | |
XM_017005707.1 | 1337 | Silent Mutation | TAC,TAT | Y,Y 312 | XP_016861196.1 | |
XM_017005708.1 | 1337 | Silent Mutation | TAC,TAT | Y,Y 312 | XP_016861197.1 | |
XM_017005709.1 | 1337 | Missense Mutation | CGC,TGC | R,C 316 | XP_016861198.1 | |
XM_017005710.1 | 1337 | Silent Mutation | TAC,TAT | Y,Y 275 | XP_016861199.1 |