Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAATATGTCACTTATATTTATAG[C/T]TGAAAAAGATTAAAATGTGAAAAAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 611916 | |||||||||||||||||||||||
Literature Links: |
COL6A5 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
COL6A5 - collagen type VI alpha 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278298.1 | Intron | NP_001265227.1 | ||||
NM_153264.6 | Intron | NP_694996.5 | ||||
XM_011512621.2 | Intron | XP_011510923.1 | ||||
XM_011512622.2 | Intron | XP_011510924.1 | ||||
XM_011512623.2 | Intron | XP_011510925.1 |