Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACCTAGATGAGCAGATTAAGAAA[A/G]TGTCCCAGCAGATCCTTGAGAAGCG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 611103 | |||||||||||||||||||||||
Literature Links: |
ACAD9 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ACAD9 - acyl-CoA dehydrogenase family member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014049.4 | 2003 | Missense Mutation | ATG,GTG | M,V 601 | NP_054768.2 | |
XM_017006264.1 | 2003 | Intron | XP_016861753.1 | |||
XM_017006265.1 | 2003 | Intron | XP_016861754.1 |
KIAA1257 - KIAA1257 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020741.2 | 2003 | Intron | NP_065792.1 | |||
XM_011513050.2 | 2003 | Intron | XP_011511352.2 | |||
XM_017006939.1 | 2003 | UTR 3 | XP_016862428.1 | |||
XM_017006940.1 | 2003 | UTR 3 | XP_016862429.1 | |||
XM_017006941.1 | 2003 | UTR 3 | XP_016862430.1 | |||
XM_017006942.1 | 2003 | UTR 3 | XP_016862431.1 | |||
XM_017006943.1 | 2003 | UTR 3 | XP_016862432.1 | |||
XM_017006944.1 | 2003 | UTR 3 | XP_016862433.1 | |||
XM_017006945.1 | 2003 | Intron | XP_016862434.1 | |||
XM_017006946.1 | 2003 | UTR 3 | XP_016862435.1 | |||
XM_017006947.1 | 2003 | UTR 3 | XP_016862436.1 | |||
XM_017006948.1 | 2003 | UTR 3 | XP_016862437.1 | |||
XM_017006949.1 | 2003 | UTR 3 | XP_016862438.1 | |||
XM_017006950.1 | 2003 | UTR 3 | XP_016862439.1 | |||
XM_017006951.1 | 2003 | UTR 3 | XP_016862440.1 | |||
XM_017006952.1 | 2003 | UTR 3 | XP_016862441.1 | |||
XM_017006953.1 | 2003 | UTR 3 | XP_016862442.1 | |||
XM_017006954.1 | 2003 | Intron | XP_016862443.1 |