Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACATATCTCAATATTCACTGAAC[A/G]AGTACATGAAACTGCCAAACTCAGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604300 | |||||||||||||||||||||||
Literature Links: |
HACL1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
HACL1 - 2-hydroxyacyl-CoA lyase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284413.1 | Intron | NP_001271342.1 | ||||
NM_001284415.1 | Intron | NP_001271344.1 | ||||
NM_001284416.1 | Intron | NP_001271345.1 | ||||
NM_012260.3 | Intron | NP_036392.2 |