Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCTGTCTCCTTAATGGTTCCTG[A/G]TGTACTGTTTTTATTTATTTGCATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GTPBP8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GTPBP8 - GTP binding protein 8 (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEPRO - nucleolus and neural progenitor protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319109.1 | 1432 | Missense Mutation | CCA,TCA | P,S 479 | NP_001306038.1 | |
NM_001319110.1 | 1432 | Missense Mutation | CCA,TCA | P,S 382 | NP_001306039.1 | |
NM_001319111.1 | 1432 | Missense Mutation | CCA,TCA | P,S 346 | NP_001306040.1 | |
NM_001319112.1 | 1432 | Missense Mutation | CCA,TCA | P,S 438 | NP_001306041.1 | |
NM_001319114.1 | 1432 | Missense Mutation | CCA,TCA | P,S 413 | NP_001306043.1 | |
NM_001319115.1 | 1432 | Missense Mutation | CCA,TCA | P,S 452 | NP_001306044.1 | |
NM_015412.3 | 1432 | Missense Mutation | CCA,TCA | P,S 549 | NP_056227.2 | |
XM_005247276.2 | 1432 | Missense Mutation | CCA,TCA | P,S 413 | XP_005247333.1 | |
XM_011512635.2 | 1432 | Missense Mutation | CCA,TCA | P,S 346 | XP_011510937.1 | |
XM_011512636.2 | 1432 | Missense Mutation | CCA,TCA | P,S 346 | XP_011510938.1 | |
XM_011512638.2 | 1432 | Missense Mutation | CCA,TCA | P,S 346 | XP_011510940.1 | |
XM_017006102.1 | 1432 | UTR 3 | XP_016861591.1 | |||
XM_017006103.1 | 1432 | UTR 3 | XP_016861592.1 |