Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAATTAAATTGTGTACTTCATCA[C/T]TGGGAAGGCACCGGAAAACCTGATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602382 | ||||||||||||||||||||
Literature Links: |
PLD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLD1 - phospholipase D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130081.2 | 3102 | Missense Mutation | AAT,AGT | N,S 969 | NP_001123553.1 | |
NM_002662.4 | 3102 | Missense Mutation | AAT,AGT | N,S 1007 | NP_002653.1 | |
XM_005247533.2 | 3102 | Missense Mutation | AAT,AGT | N,S 1007 | XP_005247590.1 | |
XM_005247534.2 | 3102 | Missense Mutation | AAT,AGT | N,S 969 | XP_005247591.1 | |
XM_011512897.1 | 3102 | Intron | XP_011511199.1 | |||
XM_011512898.1 | 3102 | Intron | XP_011511200.1 | |||
XM_011512899.2 | 3102 | Missense Mutation | AAT,AGT | N,S 590 | XP_011511201.1 | |
XM_017006623.1 | 3102 | Intron | XP_016862112.1 | |||
XM_017006624.1 | 3102 | Missense Mutation | AAT,AGT | N,S 590 | XP_016862113.1 |