Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCTCCAGTCATAGGTTGTGGTT[A/G]TCTTTGTTCTTGGTCAGAACCTGCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607010 | |||||||||||||||||||||||
Literature Links: |
DCP1A PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
DCP1A - decapping mRNA 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290204.1 | 1730 | Silent Mutation | GAC,GAT | D,D 540 | NP_001277133.1 | |
NM_001290205.1 | 1730 | Silent Mutation | GAC,GAT | D,D 506 | NP_001277134.1 | |
NM_001290206.1 | 1730 | Silent Mutation | GAC,GAT | D,D 457 | NP_001277135.1 | |
NM_001290207.1 | 1730 | Silent Mutation | GAC,GAT | D,D 419 | NP_001277136.1 | |
NM_018403.6 | 1730 | Silent Mutation | GAC,GAT | D,D 578 | NP_060873.4 |