Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGTCACCAGCAGCCACAGCCACA[C/T]GGAATGCCTCGGCCACACGGTGTTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607071 MIM: 604038 MIM: 602725 MIM: 607073 | |||||||||||||||||||||||
Literature Links: |
HYAL1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
HYAL1 - hyaluronoglucosaminidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033159.3 | 1650 | Missense Mutation | CAT,CGT | H,R 271 | NP_149349.2 | |
NM_153281.1 | 1650 | Missense Mutation | CAT,CGT | H,R 271 | NP_695013.1 | |
NM_153282.2 | 1650 | Missense Mutation | CAT,CGT | H,R 271 | NP_695014.1 | |
NM_153283.2 | 1650 | Missense Mutation | CAT,CGT | H,R 89 | NP_695015.1 | |
NM_153285.2 | 1650 | Missense Mutation | CAT,CGT | H,R 12 | NP_695017.1 | |
XM_011533667.2 | 1650 | Missense Mutation | CAT,CGT | H,R 271 | XP_011531969.1 | |
XM_011533668.2 | 1650 | Missense Mutation | CAT,CGT | H,R 271 | XP_011531970.1 | |
XM_011533669.2 | 1650 | Missense Mutation | CAT,CGT | H,R 271 | XP_011531971.1 |
HYAL3 - hyaluronoglucosaminidase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IFRD2 - interferon related developmental regulator 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAT6 - N-acetyltransferase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |