Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604038 MIM: 602725 MIM: 607073 MIM: 601281 | ||||||||||||||||||||
Literature Links: |
HYAL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HYAL3 - hyaluronoglucosaminidase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IFRD2 - interferon related developmental regulator 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LSMEM2 - leucine rich single-pass membrane protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304385.1 | 259 | Missense Mutation | AGC,GGC | S,G 58 | NP_001291314.1 | |
NM_153215.2 | 259 | Missense Mutation | ACA,GCA | T,A 58 | NP_694947.1 | |
XM_006712979.3 | 259 | Missense Mutation | ACA,GCA | T,A 67 | XP_006713042.1 | |
XM_006712980.3 | 259 | Missense Mutation | AGC,GGC | S,G 67 | XP_006713043.1 | |
XM_011533370.2 | 259 | Missense Mutation | ACA,GCA | T,A 34 | XP_011531672.1 |
NAT6 - N-acetyltransferase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEMA3B - semaphorin 3B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |